Learn More
MSC Dow Corning 3138356 3 oz Tube Clear RTV Silicone Joint Sealant -76 to 356°F Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series 732
Catalog No. 501113811
Encompass
Dow Corning 3138356 3 oz Tube Clear RTV Silicone Joint Sealant -76 to 356°F Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series 732
Catalog No. 50-111-3811 Supplier MSC Supplier No. 00231621
Dow Corning 3138356 3 oz Tube Clear RTV Silicone Joint Sealant -76 to 356°F Operating Temp, 20 min Tack Free Dry Time, 24 hr Full Cure Time, Series 732