Learn More
Ace Glass, Inc. 73mm porosity A glass filter disc designed for removable use within liquid chromatography columns with #80 Ace threads.
SDP
Supplier:  Ace Glass, Inc. 5848100
Encompass_Preferred
Catalog No. 50-972-350