Learn More
Apexbio Technology LLC 740 Y-P, 1236188-16-1, MFCD07784548, 1 mg
SDP
Supplier:  Apexbio Technology LLC B52461
Encompass_Preferred
MF: C141H222N43O39PS3, MW: 3270.72, Solubility: 163.536mg/mL in DMSO. 740 Y-P is an activator of PI3K with concentration of 20 uM.
Catalog No. 50-101-2671