Learn More
Chemglass Life Sciences Dish, Crystallizing, 740mL, 125 x 65mm
SDP
Catalog No. 501215011
Encompass_Preferred
Dish, Crystallizing, 740mL, 125 x 65mm. Tops are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization.
Catalog No. 50-121-5011 Supplier Chemglass Life Sciences Supplier No. CG8276P125
Dish, Crystallizing, 740mL, 125 x 65mm. Tops are reinforced and fire polished to reduce chipping. Will withstand repeated sterilization.