Learn More
eMolecules​ 74181-34-3 | 2,2-Dimethyl-1,3-dioxan-5-one | Combi-Blocks, Inc. | MFCD00671514 | 130.143 | C6H10O3 | 98.000 | CC1(C)OCC(=O)CO1 | 500g | 882682872
Catalog No. 5000483498 Shop All eMolecules Inc. Products
Encompass_Preferred
2,2-Dimethyl-1,3-dioxan-5-one | Combi-Blocks, Inc. | 74181-34-3 | MFCD00671514 | 130.143 | C6H10O3 | 98.000 | CC1(C)OCC(=O)CO1 | 500g | 882682872
Catalog No. 50-004-83498 Supplier eMolecules​ Supplier No. 882682872
2,2-Dimethyl-1,3-dioxan-5-one | Combi-Blocks, Inc. | 74181-34-3 | MFCD00671514 | 130.143 | C6H10O3 | 98.000 | CC1(C)OCC(=O)CO1 | 500g | 882682872