Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scott Safety™ 742 Series Air-purifying Respirator Accessory, Cartridges

Catalog No. 191501254
Change view
Click to view available options
Cartridge Type:
Acid Gas, P100
Ammonia, Methylamine
Multipurpose
Multipurpose/P100
Organic Vapor Cartridge
Organic Vapor, Acid Gas, P100
Organic Vapor, P100
Particulate, P100 Cartridge
Color:
Black/Black
Black/Magenta
Green/Green
Magenta/Magenta
Magenta/White
Magenta/Yellow
Olive/Magenta
Olive/Olive
White/White
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type Color
19-150-1254 Multipurpose/P100 Olive/Magenta
19-161-740 Multipurpose Olive/Olive
19-161-796 Particulate, P100 Cartridge Magenta/Magenta
19-161-797 Acid Gas, P100 White/White
19-161-801 Organic Vapor, P100 Black/Magenta
19-161-802 Organic Vapor, Acid Gas, P100 Magenta/Yellow
19-161-800 Organic Vapor Cartridge Black/Black
19-700-845 Acid Gas, P100 Magenta/White
19-818-556 Ammonia, Methylamine Green/Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 19-150-1254 Supplier Scott Safety™ Supplier No. 7422SD1
This item is not returnable. View return policy

Easy-breathingtwin cartridges were developed to provide optimal respiratory protection in non-IDLH industrial or HAZMAT situations

  • Low profile, swept back cartridge design ensures optimal field of vision and allows use of communications
  • Available in a full range of NIOSH approved, chemical cartridges or combination cartridges
  • Attaches directly to XCEL Halfmask and Promask full facepiece
  • Uses Quarter turn adapter when used with AV Series Facepieces

Specifications

Cartridge Type Multipurpose/P100
Certifications/Compliance NIOSH
Color Olive/Magenta
Filter Type Particulate, Acid Gases (AG), Organic Vapors (OV), Ammonia/Methylamine
Product Type Cartridge/Filter Combination
For Use With (Application) Non-IDLH and non-O2 deficient applications
For Use With (Equipment) Scott 742 Series Air Purifying Respirators
Quantity 1 Pair/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.