Learn More
eMolecules​ 7432-11-3 | 8-Azabicyclo[3.2.1]octan-3-ol | AA Blocks LLC | MFCD16664490 | 127.187 | C7H13NO | 0.000 | OC1CC2CCC(C1)N2 | 250mg | 913147408
Catalog No. NC3966479 Shop All eMolecules Inc. Products
Encompass_Preferred
8-Azabicyclo[3.2.1]octan-3-ol | AA Blocks LLC | 7432-11-3 | MFCD16664490 | 127.187 | C7H13NO | 0.000 | OC1CC2CCC(C1)N2 | 250mg | 913147408
Catalog No. NC3966479 Supplier eMolecules​ Supplier No. 913147408
8-Azabicyclo[3.2.1]octan-3-ol | AA Blocks LLC | 7432-11-3 | MFCD16664490 | 127.187 | C7H13NO | 0.000 | OC1CC2CCC(C1)N2 | 250mg | 913147408