Learn More
Hans Rudolph Inc 7450 Mask Face Piece, Small
SDP
Catalog No. NC2120634
Encompass_Preferred
7450 Series Silicone V2™ Oro-Nasal Mask is the latest face piece design from Hans Rudolph. Molded of BLUE silicone rubber, the anatomically shaped design aids in a leak free comfortable fit. New FIVE Strap adjustable headgear design. Custom mask adapters are designed and manufactured to mate most any component or flow meter. Two-Way Non-Rebreathing valves are available for this face mask. Five sizes available.
This item is not returnable. View return policy
Catalog No. NC2120634 Supplier Hans Rudolph Inc Supplier No. 669197
Customers require a valid business account associated to their profile to access pricing and ordering options.
7450 Series Silicone V2™ Oro-Nasal Mask is the latest face piece design from Hans Rudolph. Molded of BLUE silicone rubber, the anatomically shaped design aids in a leak free comfortable fit. New FIVE Strap adjustable headgear design. Custom mask adapters are designed and manufactured to mate most any component or flow meter. Two-Way Non-Rebreathing valves are available for this face mask. Five sizes available.