Learn More
CTMATTER LLC 7490016576126, SKILCRAFT Dymo LabelWriter 550 Label Printer, 62 Labels/min Print Speed, 5.34 x 8.5 x 7.38
Catalog No. NC3861262
Encompass_Preferred
Connect the label printer to your PC or Mac and you're ready to print labels, name badges and more. The SKILCRAFT Dymo LabelWriter 550 Turbo label printer comes with the unique automatic label recognition feature, so you always know the size, type, and number of labels remaining. It prints labels directly from text in Microsoft Word, Excel, Outlook, Mac address book and other popular programs. The label printer uses thermal printing technology, no expensive ink or toner needed.
Catalog No. NC3861262 Supplier CTMATTER LLC Supplier No. NSN6576126
Connect the label printer to your PC or Mac and you're ready to print labels, name badges and more. The SKILCRAFT Dymo LabelWriter 550 Turbo label printer comes with the unique automatic label recognition feature, so you always know the size, type, and number of labels remaining. It prints labels directly from text in Microsoft Word, Excel, Outlook, Mac address book and other popular programs. The label printer uses thermal printing technology, no expensive ink or toner needed.