Learn More
Enterprise Technology Solutions Dymo Digital Shipping Scale, 25 Lb Capacity, 8.2 X 9.2 Platform
SDP
Catalog No. 502041145
Encompass_Preferred
ABILITYONE 7490016881538 Skilcraft DymoÂ/SKILCRAFTÂ Digital shipping scale with LCD screen provides accurate reading of shipping weights within 0.1 ounce and includes a tare function for measuring the weight of package contents. Scale with 25-pound capacity is compatible with Windows and Macintosh operating systems and can be powered via USB cable (included) or three AAA batteries (not included).
Catalog No. 50-204-1145 Supplier Enterprise Technology Solutions Supplier No. NSN6881538
ABILITYONE 7490016881538 Skilcraft DymoÂ/SKILCRAFTÂ Digital shipping scale with LCD screen provides accurate reading of shipping weights within 0.1 ounce and includes a tare function for measuring the weight of package contents. Scale with 25-pound capacity is compatible with Windows and Macintosh operating systems and can be powered via USB cable (included) or three AAA batteries (not included).