Learn More
Med Vet International 74mm Broad Compression Plate, Six 3.5mm Holes, J0422E
Catalog No. 5000582711
Encompass_Preferred
3.5mm Compression Plate. Use with 3.5mm cortical or 3.5mm cancellous screws to produce fracture compression.
Catalog No. 50-005-82711 Supplier Med Vet International Supplier No. JOR422E
3.5mm Compression Plate. Use with 3.5mm cortical or 3.5mm cancellous screws to produce fracture compression.