Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

75/25 Methanol/Acetonitrile, Solstice

Catalog No. 60090064 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-064 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-064 Supplier Solstice Supplier No. 22214X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LabReady™ Solvent Blend

TRUSTED_SUSTAINABILITY

Specifications

CAS 67-56-1, 75-05-8
CAS Max % 75.20%, 24.80%
Quantity 4 x 4 L
Flash Point -1,1°C
Packaging Glass Bottle
Physical Form Liquid
Purity Grade Notes HPLC grade blends
Boiling Point 48.9°C
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.