Learn More
Ace Glass, Inc. 75/50 BALL JOINT (GLASSBLOWER)
SDP
Supplier:  Ace Glass, Inc. 765570
Encompass
Full length, standard wall 75/50 ball joint. This ACE ball joint is precision ground and vacuum leak-tested for fit and interchangeability.
Catalog No. 50-975-279