Learn More
Chemglass Life Sciences 75/50 O-Ring Ball Joint, Clamp Size: 75, O-Ring Size: 146, Tubing OD: 56mm
SDP
Catalog No. CG12212
Encompass_Preferred
75/50 O-Ring Ball Joint, Clamp Size: 75, O-Ring Size: 146,Tubing OD: 56mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.
Catalog No. CG12212 Supplier Chemglass Life Sciences Supplier No. CG12212
75/50 O-Ring Ball Joint, Clamp Size: 75, O-Ring Size: 146,Tubing OD: 56mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.