Learn More
Ace Glass, Inc. 75/50 SOCKET JOINT (GLASSBLOWER)
SDP
Catalog No. 50975280
Encompass
Full length, standard wall 75/50 socket joint. This ACE socket joint is precision ground and vacuum leak-tested for fit and interchangeability.
Catalog No. 50-975-280 Supplier Ace Glass, Inc. Supplier No. 765572
Full length, standard wall 75/50 socket joint. This ACE socket joint is precision ground and vacuum leak-tested for fit and interchangeability.