Learn More
Sanisure Inc Silicone Sanitary Gasket
Catalog No. NC3106265
Encompass_Preferred
SaniSure Silicone Sanitary Gaskets are designed to provide reliable, secure seals between sanitary fittings in single-use bioprocessing and pharmaceutical applications. Manufactured from platinum-cured silicone (PCS), these gaskets offer a flexible, durable sealing solution for sanitary connections. Available in multiple sizes to support a range of tubing and process requirements.
Catalog No. NC3106265 Supplier Sanisure Inc Supplier No. FTBL42RXPX075CP
SaniSure Silicone Sanitary Gaskets are designed to provide reliable, secure seals between sanitary fittings in single-use bioprocessing and pharmaceutical applications. Manufactured from platinum-cured silicone (PCS), these gaskets offer a flexible, durable sealing solution for sanitary connections. Available in multiple sizes to support a range of tubing and process requirements.