Learn More
Biolegend APC/Fire™ 750 anti-mouse IFN-γ
Supplier:  Biolegend 505860
Encompass_Preferred
APC/Fire™ 750 anti-mouse IFN-γ [XMG1.2]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: ICFC; Size: 100 ug
Catalog No. NC1770560