Learn More
Perkin Elmer US LLC 750 VA Power Conditioner for AAnalyst 100/200/300/400/PinAAcle 900 F
Supplier:  Perkin Elmer US LLC N9307521
Encompass_Preferred
Damaged or compromised components, disrupted processes, lack of reliability they all add up to frustration, broken schedules and costly downtime. The problem is caused by a host of power disturbances some visible and many invisible that threaten your equipments operation every year. Power disturbances are simply those large and small variations in the quality of the electric power you use day in and day out. Some come from your local utility company, but most are created within your own facility as a result of the distribution and use of electrical power. These variations include high-energy voltage transients, sags and swells, electrical noise, and common mode voltage, as well as the power outages everyone witnesses when the lights go out. Conditioners provide clean power Dual-output models can protect both high-voltage system loads and low-voltage peripherals at less expense and effort than installing two conditioners Protects against: high-voltage surges, common-mode voltage, e
Catalog No. 50-211-3133