Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ 6000 Series Cartridge/P100 Particulate Filter Combinations

Catalog No. 19096901 Shop All 3M Products
Change view
Click to view available options
Cartridge Type:
Acid Gas Cartridge
Ammonia, Methylamine Cartridge
Filter Combo Cartridge
Formaldehyde and Organic Vapor Cartridge
Mercury Vapor, Chlorine, Sulfur Dioxide P100
Organic Vapor Cartridge
Organic Vapor, Acid Gas Cartridge
Color:
Black/Magenta
Green/Magenta
Magenta/Black/Olive
Magenta/Orange
Magenta/White
Magenta/Yellow
Olive/Magenta
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cartridge Type Color
19-096-901 Filter Combo Cartridge Magenta/Yellow
18-999-4552 Formaldehyde and Organic Vapor Cartridge Magenta/Black/Olive
18-999-4567 Filter Combo Cartridge Olive/Magenta
18-999-4549 Acid Gas Cartridge Magenta/White
18-999-4550 Organic Vapor, Acid Gas Cartridge Magenta/Yellow
18-999-4551 Ammonia, Methylamine Cartridge Green/Magenta
18-999-4548 Organic Vapor Cartridge Black/Magenta
19-070-341 Mercury Vapor, Chlorine, Sulfur Dioxide P100 Magenta/Orange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Do not exceed maximum use concentrations established by regulatory standards. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Catalog No. 19-096-901 Supplier 3M™ Supplier No. 7000002009
Only null left
Add to Cart
Add to Cart
Do not exceed maximum use concentrations established by regulatory standards. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

Preassembled with P100 filters

  • Can be used for contaminant concentration up to 10X the Permissible Exposure Limit (PEL) with half-facepiece respirators, or 50X the PEL with full-facepiece respirators (when quantitatively fit-tested) or according to specific OSHA standards or applicable government regulations, whichever is lower
  • Not for use in atmospheres that are unknown or immediately dangerous to life or health (IDLH)
  • Cartridge/filter assemblies using P100 filters are 99.97% efficient against solid and liquid aerosols, including oils
  • 3M Catalog# 60921, 60922, 60923, 60924, 60925, 60926, 60928, 60929S

Do not exceed maximum use concentrations established by regulatory standards. These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.

TRUSTED_SUSTAINABILITY

Specifications

Cartridge Type Filter Combo Cartridge
Certifications/Compliance NIOSH
Color Magenta/Yellow
Product Type Cartridge/Filter Combination
For Use With (Application) Use against radioiodine or methyl bromide
For Use With (Equipment) 3M™ half and full facepiece 6000, 7000, ff-400 series with bayonet filter holders
Model 60928
Quantity 60/Cs.
Length (English) 4.2 in.
Length (Metric) 10.6 cm
Height (English) 3.4 in.
Height (Metric) 8.6 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.