Learn More
Alkali Scientific 75cm² Cell Culture Flasks, Vent Cap, Non-Treated, Sterile, 5/pk, 100/cs
SDP
Catalog No. 502336053
Encompass_Preferred
These Cell Culture Flasks are non-treated, vented and sterile. They come packaged in zip bags and are high clarity, 100% virgin polystyrene. They are stackable, sterilized by E-Beam, non-pyrogenic, and DNase/RNase free. They have frosted writing, clear graduations, and clear lot number for batch traceability. 100 per case.
Catalog No. 50-233-6053 Supplier Alkali Scientific Supplier No. TL32
These Cell Culture Flasks are non-treated, vented and sterile. They come packaged in zip bags and are high clarity, 100% virgin polystyrene. They are stackable, sterilized by E-Beam, non-pyrogenic, and DNase/RNase free. They have frosted writing, clear graduations, and clear lot number for batch traceability. 100 per case.