Learn More
Chemglass Life Sciences 75cm² Cell Culture Flask, Yellow Vent Cap, Non-Treated, Sterile, 5/pk, 100/cs
SDP
Catalog No. 501897660
Encompass_Preferred
Made from high clarity, 100% virgin polystyreneSterilized by E-beamClear lot number for batch traceabilityIndividually plastic wrappedNon-Pyrogenic, DNase/RNase FreeAngled neck for easy accessFrosted writing and clear graduationsNotched bottom for slip-free stackingVented caps (with breathable membrane)
Catalog No. 50-189-7660 Supplier Chemglass Life Sciences Supplier No. CGN2408750
Made from high clarity, 100% virgin polystyreneSterilized by E-beamClear lot number for batch traceabilityIndividually plastic wrappedNon-Pyrogenic, DNase/RNase FreeAngled neck for easy accessFrosted writing and clear graduationsNotched bottom for slip-free stackingVented caps (with breathable membrane)