Learn More
Bellco Glass, Inc. 75cm2 Cell Culture Flask, Vent Cap, Non-Treated, sterile 5/pk, 100/cs
SDP
Supplier:  Bellco Glass, Inc. 451400072
Encompass_Preferred
75cm2 Cell Culture Flask, Vent Cap, Non-Treated, sterile, Maximum Working Volume 250mL, 5/pk, 100/cs. cell culture flasks are available in cell growth areas ranging from 25cm2 to 225cm2. Tissue culture treated or non-treated flasks available with a vent cap or plug seal cap to meet your needs. Made of high clarity, 100% virgin polystyrene.• Sterilized by E-beam, SAL=10-6.• Non-Pyrogenic, DNase/Rnase free.• Frosted writing and clear graduations.• Notched bottom for slip free stacking.• Clear lot number for batch traceability.• Packaged in sterile, zip-sealable bags.
Catalog No. 50-232-9928