Learn More
CELLTREAT Scientific Products 75cm2 Tissue Culture Flask - Plug Seal Cap, Sterile
SDP
Catalog No. 50202079
Encompass_Preferred
CELLTREAT Scientific Products 75cm¿ Tissue Culture Treated Flask. Tissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachment. Convenient zip-closure bag is a practical alternative to folding and taping open bags. Large opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapers. Stable base enables filling with flask standing upright. Thick, durable walls. Flask is printed with lot number and expiration date for easy identification and tracking. Plug seal cap and maximum volume of 250mL. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 5/re-sealable bag, 100/case.
This item is not returnable. View return policy
Catalog No. 50-202-079 Supplier CELLTREAT Scientific Products Supplier No. 229340
CELLTREAT Scientific Products 75cm¿ Tissue Culture Treated Flask. Tissue Culture Treated Flasks have uniform hydrophilic surface ideal for consistent cell attachment. Convenient zip-closure bag is a practical alternative to folding and taping open bags. Large opening, canted neck design and moderate angles allow easy access to entire flask surface; ideal for cell scrapers. Stable base enables filling with flask standing upright. Thick, durable walls. Flask is printed with lot number and expiration date for easy identification and tracking. Plug seal cap and maximum volume of 250mL. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, 5/re-sealable bag, 100/case.