Learn More
The Fresh Test 75g Glucose Bottle - Plastic 1/ea, Non-GMO Dextrose
Catalog No. NC3362997 Shop All The Fresh Test Diagnostic Glucose Test Products
Encompass
The Fresh Test 75g Glucose Bottle - Plastic is a diagnostic glucose beverage used in the screening of Diabetes Mellitus and Gestational Diabetes Mellitus. Each bottle contains 75 grams of glucose in powder form. Patients should add water to the fill line, shake well, and consume within 5 minutes for a Glucose Load Test.
Ingredients / Certifications
Non-GMO Dextrose, Crystallized Lemon, Organic Mint
30-month shelf life
BPA-free bottle
Kosher Certified; Non-GMO Project Verified
Organic Ingredients; Gluten-Free; Dye-Free; Additive-Free; Preservative-Free; Sulfite-Free
The Glucose Bottle - Plastic is Fresh Test's most popular product, trusted by laboratories and preferred by patients for its taste and ease of use.
Manufacturer Information
Fresh Test, LLC is a Class II Medical Device manufacturer of diagnostic and laboratory supplies.
This item is not returnable. View return policy
Catalog No. NC3362997 Supplier The Fresh Test Supplier No. 27102058PB
The Fresh Test 75g Glucose Bottle - Plastic is a diagnostic glucose beverage used in the screening of Diabetes Mellitus and Gestational Diabetes Mellitus. Each bottle contains 75 grams of glucose in powder form. Patients should add water to the fill line, shake well, and consume within 5 minutes for a Glucose Load Test.
Ingredients / Certifications
Non-GMO Dextrose, Crystallized Lemon, Organic Mint
30-month shelf life
BPA-free bottle
Kosher Certified; Non-GMO Project Verified
Organic Ingredients; Gluten-Free; Dye-Free; Additive-Free; Preservative-Free; Sulfite-Free
The Glucose Bottle - Plastic is Fresh Test's most popular product, trusted by laboratories and preferred by patients for its taste and ease of use.
Manufacturer Information
Fresh Test, LLC is a Class II Medical Device manufacturer of diagnostic and laboratory supplies.