Learn More
The Fresh Test 75g Glucose Powder 1/ea, Non-GMO Dextrose, Crystallized Lemon, Organic Mint, 30-month shelf life, BPA-free pouch, Kosher Certified, Non-GMO Project Verified, Organic Ingredients, Gluten-Free, Dye-Free, Additive-Free, Preservative-Free, Sulfite-Free
Catalog No. NC3362994 Shop All The Fresh Test Diagnostic Glucose Test Products
Encompass
The Fresh Test 75g Glucose Powder is a diagnostic glucose beverage intended for use in the screening of Diabetes Mellitus and Gestational Diabetes Mellitus. Each pouch contains 75 grams of glucose to be reconstituted with 10 oz. of water prior to administration. For an optimal experience, the beverage should be consumed cold. When used for a 75-gram glucose load test, the beverage should be consumed within 5 minutes, followed by a plasma glucose measurement at 1 hour.
Ingredients / Certifications
Non-GMO Dextrose, Crystallized Lemon, Organic Mint
30-month shelf life
BPA-free pouch
Kosher Certified; Non-GMO Project Verified
Organic Ingredients; Gluten-Free; Dye-Free; Additive-Free; Preservative-Free; Sulfite-Free
Manufacturer Information
Fresh Test, LLC is a Class II Medical Device manufacturer of diagnostic and laboratory supplies.
This item is not returnable. View return policy
Catalog No. NC3362994 Supplier The Fresh Test Supplier No. 27102056
The Fresh Test 75g Glucose Powder is a diagnostic glucose beverage intended for use in the screening of Diabetes Mellitus and Gestational Diabetes Mellitus. Each pouch contains 75 grams of glucose to be reconstituted with 10 oz. of water prior to administration. For an optimal experience, the beverage should be consumed cold. When used for a 75-gram glucose load test, the beverage should be consumed within 5 minutes, followed by a plasma glucose measurement at 1 hour.
Ingredients / Certifications
Non-GMO Dextrose, Crystallized Lemon, Organic Mint
30-month shelf life
BPA-free pouch
Kosher Certified; Non-GMO Project Verified
Organic Ingredients; Gluten-Free; Dye-Free; Additive-Free; Preservative-Free; Sulfite-Free
Manufacturer Information
Fresh Test, LLC is a Class II Medical Device manufacturer of diagnostic and laboratory supplies.