Learn More
Eagle Stainless Container Eagle Stainless, 75 Liter Slant Bottom Container with 1.5" Tri-clamp outlet. 316L Stainless Steel. (KTT-CTL Series)
SDP
Catalog No. NC1471584
Encompass_Preferred
60 Liter slant bottom container with 1.5" Tri-clamp outlet. Supplied with lid, platinum cured silicone gasket and lock lever ring. Type 316L stainless steel. Pharmaceutical grade finish of 10 Ra or better.
Catalog No. NC1471584 Supplier Eagle Stainless Container Supplier No. KTTCTL47316LJ
60 Liter slant bottom container with 1.5" Tri-clamp outlet. Supplied with lid, platinum cured silicone gasket and lock lever ring. Type 316L stainless steel. Pharmaceutical grade finish of 10 Ra or better.