Learn More
Ace Glass, Inc. 75MM 24/40 FUNNEL 75MM 24/40 FUNNEL
SDP
Catalog No. 50974707
Encompass_Preferred
75mm powder funnel with 24/40 inner joint is useful in pouring powders or liquids into ground joint containers.
Catalog No. 50-974-707 Supplier Ace Glass, Inc. Supplier No. 723608
Add to Cart
Add to Cart
75mm powder funnel with 24/40 inner joint is useful in pouring powders or liquids into ground joint containers.