Learn More
Chemglass Life Sciences Filter Funnel, Hirsh, 75mm Diameter Funnel Top, 30mm Coarse Frit
SDP
Catalog No. CG140806
Encompass_Preferred
Filter Funnel, Hirsh, 75mm Diameter Funnel Top, 30mm Coarse Frit. Used in applications where it is necessary to wash and re-dissolve the precipitate with chemicals that would attack standard filter paper. The 60º angle on the funnel permits removal of precipitates.
Catalog No. CG140806 Supplier Chemglass Life Sciences Supplier No. CG140806
Add to Cart
Add to Cart
Filter Funnel, Hirsh, 75mm Diameter Funnel Top, 30mm Coarse Frit. Used in applications where it is necessary to wash and re-dissolve the precipitate with chemicals that would attack standard filter paper. The 60º angle on the funnel permits removal of precipitates.