Learn More
Ace Glass, Inc. 75MM JOINTS (GB)
SDP
Catalog No. 50975238
Encompass_Preferred
75mm glass o-ring seal joint tooled with a groove to take a -341 FETFE o-ring, and to provide a greaseless, vacuum-tight connection. Supplied with one o-ring, which may be used up to 230°C.
Catalog No. 50-975-238 Supplier Ace Glass, Inc. Supplier No. 764618
75mm glass o-ring seal joint tooled with a groove to take a -341 FETFE o-ring, and to provide a greaseless, vacuum-tight connection. Supplied with one o-ring, which may be used up to 230°C.