Learn More
Ace Glass, Inc. Fritted disc, 75mm OD, porosity C (25-50 micron), fits3708-02, 3709-05
SDP
Supplier:  Ace Glass, Inc. 370375
Encompass_Preferred
75mm O.D. Porosity C (25-50 micron) fritted disc.
Catalog No. 50-971-349