Learn More
Bulbs Com Philips 75W 62in T5 TUV Germicidal Fluorescent Tube (Pack of 4)
SDP
Catalog No. NC1753123
Encompass
Philips Sterilamp brand T5 germicidal fluorescent lamp. 75 Watts and 62" for use in germicidal sterilization applications. Produces UVC radiation to kill microscopic organisms. CAUTION: can be very harmful to eyes and skin. DO NOT expose eyes or unprotected skin to these lamps. SERIOUS injury and blindness could result. Bulb has both professional water and air disinfection application. The small 16 mm diameter of the lamp allows for a small system design and design flexibility. TUV T5 lamps offer constant UV output over their complete lifetime, for maximum security of disinfection and high system efficacy.
Catalog No. NC1753123 Supplier Bulbs Com Supplier No. TUV64T5SP
Philips Sterilamp brand T5 germicidal fluorescent lamp. 75 Watts and 62" for use in germicidal sterilization applications. Produces UVC radiation to kill microscopic organisms. CAUTION: can be very harmful to eyes and skin. DO NOT expose eyes or unprotected skin to these lamps. SERIOUS injury and blindness could result. Bulb has both professional water and air disinfection application. The small 16 mm diameter of the lamp allows for a small system design and design flexibility. TUV T5 lamps offer constant UV output over their complete lifetime, for maximum security of disinfection and high system efficacy.