Learn More
Electron Microscopy Sciences Large Glass Slide 75 x 38 mm, 1 mm thick
SDP
Catalog No. 50366186
Encompass_Preferred
In addition to our comprehensive collection of glass slides manufactured by ESCO we now carry large glass slides. These slides are precleaned, individually selected and are suitable for large specimen sections, such as, brain tissue.
Size: 75 x 38 mm
Thickness: 1.0 mm
Catalog No. 50-366-186 Supplier Electron Microscopy Sciences Supplier No. 7186001
In addition to our comprehensive collection of glass slides manufactured by ESCO we now carry large glass slides. These slides are precleaned, individually selected and are suitable for large specimen sections, such as, brain tissue.
Size: 75 x 38 mm
Thickness: 1.0 mm