Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS Poretics™ 76 mm Polycarbonate Disc

Catalog No. 09735210 Shop All GVS Products
Change view
Click to view available options
Pore Size:
0.1 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
09-735-210 0.1 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-735-210 Supplier GVS Poretics™ Supplier No. 1220970
Add to Cart
Add to Cart

Made from a thin, microporous polycarbonate film material. It is ideally suited for use in blood assays and high-purity and general filtration. Though polycarbonate is hydrophobic, the membrane is treated with a wetting agent to make it hydrophilic.

Properties

  • The polycarbonate membrane is manufactured by GVS using a two-stage production process that applies high quality standards.
  • This unique process allows greater control over pore size and density to ensure that the physical properties of each membrane precisely match your specifications.
  • The resulting membrane is a thin, translucent, microporous polycarbonate film with a smooth, flat surface. All particles larger than pore size are captured on its surface. It is ideal for use when collecting samples for blood tests or for high purity, general purpose filtration.

Characteristics

  • Absolute pore size and density provides flow control for liquids moving through the membrane capturing 100 percent of cells larger than pore size.
  • Smooth, thin, glass-like surface planar surface makes it ideal for particle identification by microscopy.
  • Superior strength tensile strength of 207 bar (> 3000 psi) maintains pore size and density, and will not stretch.
  • Low extractables ensure tests will be clean promoting consistent results.
  • Negligible absorption/adsorption maximizes critical solution recovery.
  • Available as hydrophilic or hydrophobic allows for a wide range of product applications.

Applications

  • Flow control of reagents through assay
  • General filtration
  • Precise filtration and prefiltration
  • Removal of red blood cells from plasma

Specifications

Autoclavable Not Autoclavable
Disposable Limited-use
Filter Type Membrane Filter
Format Disc
Material PCTE
Pore Size 0.1 μm
Sterility Non-sterile
Type Disc Membrane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.