Learn More
Cole-Parmer Jenway 7615 Scanning UV/Visible Spectrophotometer, 198 to 1000 nm, White ABS Plastic; 100-240 VAC, 50/60 Hz
Catalog No. 501961217 Shop All Cole-Parmer Products
Encompass_Preferred
Designed with your convenience in mind, the 74 series of spectrophotometers provides accurate and reliable results in various applications from teaching, to industrial applications, to routine sample analysis in quality control environments. Navigation is fast and intuitive on the large 7 high-definition color touchscreen display. View scans and curves, zoom in and out, and select points right on the screen even when wearing gloves. The home screen provides easy access to measurement modes as well as saved results and methods. Programmed measurement modes include photometric for single wavelength measurement, concentration for measuring concentration using a standard or factor, spectrum for a high purity scan across the entire wavelength range, quantitation with the ability to create a calibration curve using up to 20 standards, kinetics to monitor change over time, as well as multi-wavelength mode. Connect to a PC via the Ethernet connection or multiple USB ports.
Catalog No. 50-196-1217 Supplier Cole-Parmer Supplier No. 8305626
Add to Cart
Add to Cart
Request a Quote
Designed with your convenience in mind, the 74 series of spectrophotometers provides accurate and reliable results in various applications from teaching, to industrial applications, to routine sample analysis in quality control environments. Navigation is fast and intuitive on the large 7 high-definition color touchscreen display. View scans and curves, zoom in and out, and select points right on the screen even when wearing gloves. The home screen provides easy access to measurement modes as well as saved results and methods. Programmed measurement modes include photometric for single wavelength measurement, concentration for measuring concentration using a standard or factor, spectrum for a high purity scan across the entire wavelength range, quantitation with the ability to create a calibration curve using up to 20 standards, kinetics to monitor change over time, as well as multi-wavelength mode. Connect to a PC via the Ethernet connection or multiple USB ports.