Learn More
Hillrom 767-Series Aneroid Sphygmomanometer
Catalog No. 22355143
Change view
Click to view available options
Includes:
Five-Leg Mobile Stand
Wall Mounting
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-355-143 Wall Mounting
22-355-144 Five-Leg Mobile Stand
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-355-143 Supplier Hillrom Supplier No. WEL 767001
 Restricted Item
Add to Cart
Add to Cart
This item is not returnable. View return policy
The versatile 767 Wall and Mobile Aneroid, with improved ergonomics and a new modern design, is the smart choice for accurate and reliable readings throughout your facility.
  • Lifetime calibration warranty reduces cost of ownership
  • New 5-leg stand provides improved stability and maneuverability
  • Certified accuracy to ±3mmHg
  • Latex- and Mercury-free
Specifications
Size 11 Adult
For Use With (Application) Accurate and Reliable Blood Pressure Readings
Includes Wall Mounting
Product Type Aneroid Sphygmomanometer