Learn More
Ace Glass, Inc. Acrylic safety shield and aluminum safety cap for Michel-Miller chromatography columns. Cap is 76mm ID and fits#50 threaded columns. The shield is 60cm long.
SDP
Catalog No. 5097272
Encompass_Preferred
Catalog No. 50-972-72 Supplier Ace Glass, Inc. Supplier No. 579864