Learn More
Ace Glass, Inc. Acrylic safety shield and polyethylene safety cap for Michel-Miller chromatography columns. Cap is 76mm ID and fits#50 threaded columns. The shield is 60cm long.
SDP
Catalog No. 5097287
Encompass_Preferred
Catalog No. 50-972-87 Supplier Ace Glass, Inc. Supplier No. 5798264