Learn More
Ace Glass, Inc. PTFE multi-paddle agitator, 76mm diameter
SDP
Catalog No. 509558267
Encompass_Preferred
76mm diameter PTFE multi-paddle agitator for 10mm OD drilled or knob type shafts. Paddles are replaceable.
Catalog No. 50-955-8267 Supplier Ace Glass, Inc. Supplier No. 808908
76mm diameter PTFE multi-paddle agitator for 10mm OD drilled or knob type shafts. Paddles are replaceable.