Learn More
Ace Glass, Inc. 76mm ID 60cm long acrylic safety shield without the use with Michel-Miller chromatography columns.
SDP
Supplier:  Ace Glass, Inc. 579862
Encompass_Preferred
76mm I.D. x 60cm L Acrylic Safety Shield without cap, for use with Michel-Miller chromatography columns. Safety shield insures safe operation at pressures to 300 psi. Shield wall thickness: 3mm. The top thread of the column is hung in a polyethylene cap; the cap is then held by the plastic shield. The shield should be secured with a clamp.
Catalog No. 50-978-430