Learn More
Ace Glass, Inc. 76mm ID polyethylene shield cap fitting #50 threaded chromatography columns.
SDP
Catalog No. 50978432
Encompass_Preferred
76mm I.D. Polyethylene Shield Cap, for use with acrylic safety shield for #50 threaded Michel-Miller Chromatography Columns. Safety shield insures safe operation at pressures to 300 psi.
Catalog No. 50-978-432 Supplier Ace Glass, Inc. Supplier No. 5798144
76mm I.D. Polyethylene Shield Cap, for use with acrylic safety shield for #50 threaded Michel-Miller Chromatography Columns. Safety shield insures safe operation at pressures to 300 psi.