Learn More
M2 Polymer Technologies Inc 770-WASTELOCK-ABSORBANT 50 LB
SDP
Catalog No. NC9651533
Encompass_Preferred
A 50 lbs bag of a cross-linked Sodium Polyacryalte Superabsorbent polymer. Product absorbs as much as 500 times its weight in DI water. Particle size 200 - 800 um.
Catalog No. NC9651533 Supplier M2 Polymer Technologies Inc Supplier No. 77050LB BAG
A 50 lbs bag of a cross-linked Sodium Polyacryalte Superabsorbent polymer. Product absorbs as much as 500 times its weight in DI water. Particle size 200 - 800 um.