Learn More
MSC Loctite 235028 8 oz Can High Temperature Anti-Seize Lubricant Nickel, -54 to 2,399°F, Silver Colored, Water Resistant
Catalog No. 501112916
Encompass
Loctite 235028 8 oz Can High Temperature Anti-Seize Lubricant Nickel, -54 to 2,399°F, Silver Colored, Water Resistant
Catalog No. 50-111-2916 Supplier MSC Supplier No. 00270207
Loctite 235028 8 oz Can High Temperature Anti-Seize Lubricant Nickel, -54 to 2,399°F, Silver Colored, Water Resistant