Learn More
Waters Corp Waters Performance Maintenance Kit, For Rheodyne 7750E-075 Injector, PEEK
Supplier:  Waters Corp 201000125
Encompass_Preferred
This Performance Maintenance Kit is designed for Rheodyne 7750E-075 Injector Valves to ensure optimal operation. It provides essential components for routine upkeep.
  • Includes one rotor seal made of PEEK.
  • Contains one stator face assembly made of PEEK.
  • Features one isolation seal.
  • Comes with one hex key (9/64 in).
  • Instructions are included for easy maintenance.
  • Compatible with 7750E-075 Injector Valves.
Catalog No. 50-791-554