Learn More
eMolecules​ 7752-78-5 | 5-Bromo-2-methylpyrimidine | Apollo Scientific US | MFCD07375143 | 173.013 | C5H5BrN2 | 95.000 | Cc1ncc(Br)cn1 | 25g | 870071508
Catalog No. 5000492911 Shop All eMolecules Inc. Products
Encompass_Preferred
5-Bromo-2-methylpyrimidine | Apollo Scientific US | 7752-78-5 | MFCD07375143 | 173.013 | C5H5BrN2 | 95.000 | Cc1ncc(Br)cn1 | 25g | 870071508
Catalog No. 50-004-92911 Supplier eMolecules​ Supplier No. 870071508
5-Bromo-2-methylpyrimidine | Apollo Scientific US | 7752-78-5 | MFCD07375143 | 173.013 | C5H5BrN2 | 95.000 | Cc1ncc(Br)cn1 | 25g | 870071508