Learn More
ADMINMED NANOBIOSCIENCES 777 Microneedle Liquid Injection System
SDP
Catalog No. NC2292704
Encompass
AdminPen 777 microneedle liquid injection system is suitable for attachment to a standard syringe. The system has one hundred twenty one 700 um-tall microneedles on 1cm2 circular microneedle array. The microneedle array is made of SS316L stainless steel.
This item is not returnable. View return policy
Catalog No. NC2292704 Supplier ADMINMED NANOBIOSCIENCES Supplier No. 405
AdminPen 777 microneedle liquid injection system is suitable for attachment to a standard syringe. The system has one hundred twenty one 700 um-tall microneedles on 1cm2 circular microneedle array. The microneedle array is made of SS316L stainless steel.