Learn More
Newcomer Supply Slide Draining Tray, Polyethylene, 78 Slide Capacity
SDP
Catalog No. NC0582944
Encompass
The Slide Draining Tray is made from chemical-resistant polyethylene and can hold up to 78 slides. It has rubber feet for bench top stability. Also available in 15 and 40 slide capacity.
Catalog No. NC0582944 Supplier Newcomer Supply Supplier No. 6842D
The Slide Draining Tray is made from chemical-resistant polyethylene and can hold up to 78 slides. It has rubber feet for bench top stability. Also available in 15 and 40 slide capacity.