Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hughes 79-Gallon Temperature Controlled Safety Shower and Eye/Face Wash

Catalog No. 18666704 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Application):
C1D2 Hazardous Areas
Non-hazardous Areas
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
18-666-704 Non-hazardous Areas
18-666-703 C1D2 Hazardous Areas
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-666-704 Supplier Justrite Manufacturing Co Supplier No. TC100KS11K2

Hughes 79-Gallon Temperature Crontrolled Safety Shower and Eye/Face Wash is for outdoor use in low ambient temperatures and ensures delivery of tepid water when connected to water source

Features:

  • Delivery of tepid water for the ANSI recommended minimum period of 15 minutes
  • 79 US gallon (300 liter) tank with 3kW, 240V heater
  • Overhead shower and front mounted eye/face wash with protective lid are trace tape heated to prevent freezing.
  • Specially designed, high-performance quarter-turn ball valve made of 316L stainless steel endures the harshest environments and site conditions.
  • Low operating torque to open valve offers reliable, easy on/off operation when emergencies occur.
  • Valve includes an integral, heavy-duty stainless steel stop-plate to handle rough operation without failure.
  • Durable, coated stainless steel shower pull handle with nylon valve connection is easy to grip and activate for immediate water drenching.
  • Removable, self-draining overhead nozzle designed for high flow rates
  • Durable nylon nozzle offers optimal spray distribution for user protection.
  • Corrosion-resistant fiberglass eye/face wash incorporates an integral lid connected to the water valve for automatic operation—activate by simply lifting the lid.
  • Includes an inspection record tag for recording service and test history.
  • Electrical supply: 240V Single Phase, 60/50Hz
  • High flow rate thermostatically controlled mixing valve maintains water at a constant temperature between 68 and 86°F (20 and 30°C)
  • Immersion heated hot water tank is pre-insulated with polyurethane foam to minimize heat loss
  • Use in freezing conditions with a minimum ambient operating temperature of -4°F (-20°C) or greater
  • Insulated stainless steel frame and pipes with fiberglass outer casing provide improved corrosion resistance

Compliance

ANSI Z358.1-2014 and EN15154 Compliant

Warranty and Services

1 Year

Specifications

Activation Hand Lever: Lid Lift
Bowl Size (Metric) 356 mm
Certifications/Compliance ANSI Z358.1-2014 and EN15154 Compliant
Color White with Green Stripes
Depth (English) 115 in.
Depth (Metric) 2921 mm
Flow Rate 5.3 gpm
For Use With (Application) Non-hazardous Areas
Height (English) 76 in.
Includes 1 Unit
Inlet Connection (Metric) 1.25 in. (F) NPT
Material Polyurethane Foam, MDPE, Stainless Steel, GRP Composite, Nylon 6
Material (Piping Connection) Stainless Steel
Mounting Floor
Pressure 32 to 87 psi
Product Type Safety Shower and Emergency Eye Wash
Width (English) 53 in.
Width (Metric) 1346 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.