Learn More
Hampton Research HR2-406-66/Detergent Screen #66 (F6), 0.5 ml - 790.0 mM MEGA-8
SDP
Catalog No. 500703
Encompass
HR2-406-66/Detergent Screen #66 (F6), 0.5 ml - 790.0 mM MEGA-8. Please note items from this supplier are non-returnable.
Catalog No. 50-070-3 Supplier Hampton Research Supplier No. HR240666
HR2-406-66/Detergent Screen #66 (F6), 0.5 ml - 790.0 mM MEGA-8. Please note items from this supplier are non-returnable.