Learn More
Avanti Polar Lipids 1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (nickel salt), 231615-77-3, MFCD02259231, 25mg
Catalog No. NC1279524
Encompass
Molecular Formula: C53H93N2O15Ni, Formula Weight: 1057.003, Storage Temperature: -20°C.
Catalog No. NC1279524 Supplier Avanti Polar Lipids Supplier No. 790404C25MG
Molecular Formula: C53H93N2O15Ni, Formula Weight: 1057.003, Storage Temperature: -20°C.