Learn More
Enterprise Technology Solutions Floor Pads, 14" X 20", Maroon, 10/Box
SDP
Catalog No. 502041055
Encompass_Preferred
ABILITYONE 7910016742654 Skilcraft Ideal for resilient tile, terrazzo, marble and ceramic floors. Used on concentrated weight, rotary disk floor machines rated between 150-350 rpm. Pads are washable, reusable and corrosion-resistant.
Catalog No. 50-204-1055 Supplier Enterprise Technology Solutions Supplier No. NSN6742654
ABILITYONE 7910016742654 Skilcraft Ideal for resilient tile, terrazzo, marble and ceramic floors. Used on concentrated weight, rotary disk floor machines rated between 150-350 rpm. Pads are washable, reusable and corrosion-resistant.